Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCCTGCCACACCAGGAAACTCAC[A/G]CGGACAGCGGCCTGGCACGCAGCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604262 MIM: 612689 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
APBA3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
APBA3 - amyloid beta precursor protein binding family A member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004886.3 | 1594 | Silent Mutation | CGC,CGT | R,R 465 | NP_004877.1 | |
XM_006722950.3 | 1594 | Silent Mutation | CGC,CGT | R,R 465 | XP_006723013.1 | |
XM_006722951.3 | 1594 | Silent Mutation | CGC,CGT | R,R 223 | XP_006723014.1 |
TJP3 - tight junction protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |