Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGCTAATTTTTGTGTTTTTTTGT[C/T]TTGTTGGTAGAGATGAGGTTTCACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605546 | ||||||||||||||||||||
Literature Links: |
GP6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GP6 - glycoprotein VI platelet | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083899.2 | 1681 | Missense Mutation | AAC,GAC | N,D 553 | NP_001077368.2 | |
NM_001256017.2 | 1681 | UTR 3 | NP_001242946.2 | |||
NM_016363.5 | 1681 | UTR 3 | NP_057447.5 |