Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTAGTAAGGTTTGCAGATTGGTT[A/G]AAAGCTTTGTCACATCTCTCACATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF675 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF675 - zinc finger protein 675 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138330.2 | 2435 | Intron | NP_612203.2 | |||
XM_017026442.1 | 2435 | Silent Mutation | TTC,TTT | F,F 469 | XP_016881931.1 | |
XM_017026443.1 | 2435 | Silent Mutation | TTC,TTT | F,F 469 | XP_016881932.1 | |
XM_017026444.1 | 2435 | Intron | XP_016881933.1 |