Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCTCTGCGCTGCTGCACGCCAAC[A/C]ACACACTCAGCACCATTGACCACCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605784 | ||||||||||||||||||||
Literature Links: |
TTYH1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TTYH1 - tweety family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005367.2 | 513 | Missense Mutation | AAC,CAC | N,H 131 | NP_001005367.1 | |
NM_001201461.1 | 513 | Missense Mutation | AAC,CAC | N,H 131 | NP_001188390.1 | |
NM_020659.3 | 513 | Missense Mutation | AAC,CAC | N,H 131 | NP_065710.1 | |
XM_017027025.1 | 513 | Missense Mutation | AAC,CAC | N,H 46 | XP_016882514.1 | |
XM_017027026.1 | 513 | Missense Mutation | AAC,CAC | N,H 46 | XP_016882515.1 |