Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTACGTGGAGGTGATGCTGGAGACC[C/T]GTGGGCTTCTGGTCGCACTGGGTAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604755 | ||||||||||||||||||||
Literature Links: |
LOC105372476 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC105372476 - uncharacterized LOC105372476 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF460 - zinc finger protein 460 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006635.3 | 273 | Missense Mutation | CGT,TGT | R,C 46 | NP_006626.3 | |
XM_005258422.3 | 273 | Missense Mutation | CGT,TGT | R,C 5 | XP_005258479.1 | |
XM_017026176.1 | 273 | Missense Mutation | CGT,TGT | R,C 39 | XP_016881665.1 |