Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTGAATCTGATCCTCCCAGGTCC[A/G]TCGGAAGTTAGAGCTGGAAGCTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613432 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HAUS5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HAUS5 - HAUS augmin like complex subunit 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015302.1 | 162 | Missense Mutation | CAT,CGT | H,R 75 | NP_056117.1 | |
XM_011526684.2 | 162 | UTR 5 | XP_011524986.1 |