Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTCACCTCCTCCTTGGTCATGAC[A/G]ACGTGCTCAGGGACTAGCTGCCGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601155 MIM: 180664 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP45 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGAP45 - Rho GTPase activating protein 45 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POLR2E - RNA polymerase II subunit E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316323.1 | 338 | Silent Mutation | GTC,GTT | V,V 56 | NP_001303252.1 | |
NM_001316324.1 | 338 | Silent Mutation | GTC,GTT | V,V 56 | NP_001303253.1 | |
NM_002695.3 | 338 | Silent Mutation | GTC,GTT | V,V 149 | NP_002686.2 | |
XM_011528070.2 | 338 | Silent Mutation | GTC,GTT | V,V 149 | XP_011526372.1 | |
XM_017026883.1 | 338 | Silent Mutation | GTC,GTT | V,V 56 | XP_016882372.1 |