Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGTGAGGCCCCAGAGTGGCAGGG[G/T]CATCAGGAGGCGGGAGGGGTGGACG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604556 MIM: 134795 | ||||||||||||||||||||
Literature Links: |
DYRK1B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DYRK1B - dual specificity tyrosine phosphorylation regulated kinase 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004714.2 | 2051 | Missense Mutation | GAC,GCC | D,A 608 | NP_004705.1 | |
NM_006483.2 | 2051 | Missense Mutation | GAC,GCC | D,A 568 | NP_006474.1 | |
NM_006484.2 | 2051 | Missense Mutation | GAC,GCC | D,A 580 | NP_006475.1 | |
XM_005259398.4 | 2051 | Missense Mutation | GAC,GCC | D,A 608 | XP_005259455.1 |
FBL - fibrillarin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6719 - microRNA 6719 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |