Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTTACTGCAGGCTGGCTGAAGGC[A/G]GTGCAGGTCCTCATGGTGCTCTCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615038 MIM: 602335 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC114 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC114 - coiled-coil domain containing 114 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EMP3 - epithelial membrane protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001313905.1 | 380 | Silent Mutation | GCA,GCG | A,A 65 | NP_001300834.1 | |
NM_001425.2 | 380 | Silent Mutation | GCA,GCG | A,A 65 | NP_001416.1 | |
XM_011526605.2 | 380 | Silent Mutation | GCA,GCG | A,A 65 | XP_011524907.1 |
TMEM143 - transmembrane protein 143 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |