Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTATAGGGCCCGGCACCTGGGACC[A/G]TGTGTCCGAGGCTGGGGCTTGGGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601480 | ||||||||||||||||||||
Literature Links: |
MYO1F PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MYO1F - myosin IF | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012335.3 | 3192 | Silent Mutation | CAC,CAT | H,H 1042 | NP_036467.2 | |
XM_011528024.1 | 3192 | Silent Mutation | CAC,CAT | H,H 1063 | XP_011526326.1 | |
XM_011528025.1 | 3192 | Silent Mutation | CAC,CAT | H,H 1059 | XP_011526327.1 | |
XM_011528026.1 | 3192 | Silent Mutation | CAC,CAT | H,H 1034 | XP_011526328.1 | |
XM_011528027.1 | 3192 | Silent Mutation | CAC,CAT | H,H 1018 | XP_011526329.1 | |
XM_011528028.2 | 3192 | Intron | XP_011526330.1 | |||
XM_017026821.1 | 3192 | Silent Mutation | CAC,CAT | H,H 1038 | XP_016882310.1 |
ZNF414 - zinc finger protein 414 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |