Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCATGGACCTGCCCGAGGGCCCG[A/G]TGGGTGGCCCCACTGCGGAAAGTAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CCDC155 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC155 - coiled-coil domain containing 155 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144688.4 | 227 | Missense Mutation | ATG,GTG | M,V 8 | NP_653289.3 | |
XM_005258549.3 | 227 | Missense Mutation | ATG,GTG | M,V 8 | XP_005258606.1 | |
XM_011526488.2 | 227 | Missense Mutation | ATG,GTG | M,V 8 | XP_011524790.1 | |
XM_011526489.2 | 227 | Missense Mutation | ATG,GTG | M,V 8 | XP_011524791.1 | |
XM_011526490.2 | 227 | Missense Mutation | ATG,GTG | M,V 8 | XP_011524792.1 | |
XM_011526494.2 | 227 | UTR 5 | XP_011524796.1 | |||
XM_011526495.2 | 227 | UTR 5 | XP_011524797.1 | |||
XM_011526496.2 | 227 | Intron | XP_011524798.1 | |||
XM_011526497.2 | 227 | Missense Mutation | ATG,GTG | M,V 8 | XP_011524799.1 | |
XM_017026333.1 | 227 | Intron | XP_016881822.1 |
LOC101928295 - uncharacterized LOC101928295 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |