Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCACCTTGTGGAAGTGTAGGACAG[C/T]GTAGTGGAGCTCCTGCTCATCTTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604405 | ||||||||||||||||||||
Literature Links: |
SIGLEC6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SIGLEC6 - sialic acid binding Ig like lectin 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011527533.2 | 1359 | Missense Mutation | XP_011525835.1 | |||
XM_011527534.2 | 1359 | UTR 3 | XP_011525836.1 | |||
XM_011527535.2 | 1359 | Missense Mutation | XP_011525837.1 | |||
XM_011527536.2 | 1359 | Missense Mutation | XP_011525838.1 | |||
XM_011527537.2 | 1359 | Missense Mutation | XP_011525839.1 | |||
XM_011527538.2 | 1359 | UTR 3 | XP_011525840.1 | |||
XM_011527539.2 | 1359 | UTR 3 | XP_011525841.1 | |||
XM_017027511.1 | 1359 | Intron | XP_016883000.1 |