Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCCAGGGTGGAGAGTTGCAGCAG[C/T]GGGGGCCGCAGAGTGCCCAGCTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 151440 MIM: 164005 MIM: 611669 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LYL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LYL1 - LYL1, basic helix-loop-helix family member | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005583.4 | 1465 | Silent Mutation | CCA,CCG | P,P 86 | NP_005574.2 | |
XM_005259912.3 | 1465 | Silent Mutation | CCA,CCG | P,P 12 | XP_005259969.1 | |
XM_017026816.1 | 1465 | Silent Mutation | CCA,CCG | P,P 157 | XP_016882305.1 | |
XM_017026817.1 | 1465 | Silent Mutation | CCA,CCG | P,P 108 | XP_016882306.1 |
NFIX - nuclear factor I X | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRMT1 - tRNA methyltransferase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |