Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACAGGGCTGGTAGTTCTTCACCAG[C/T]GGCTGGAAGGCTTTCCAGAGGGCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 126350 MIM: 600599 MIM: 612256 | ||||||||||||||||||||
Literature Links: |
DNASE2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DNASE2 - deoxyribonuclease II, lysosomal | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001375.2 | 1163 | Silent Mutation | NP_001366.1 |
KLF1 - Kruppel like factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAST1 - microtubule associated serine/threonine kinase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |