Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCACTTGTTCCCGGAGCTTCCGGT[C/G]TTTCAGCCGCACCTGGCGGAACCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602754 | ||||||||||||||||||||
Literature Links: |
KCNN4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KCNN4 - potassium calcium-activated channel subfamily N member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002250.2 | 1468 | Missense Mutation | CAC,GAC | H,D 358 | NP_002241.1 | |
XM_005258882.2 | 1468 | Missense Mutation | CAC,GAC | H,D 326 | XP_005258939.1 | |
XM_005258883.2 | 1468 | Missense Mutation | CAC,GAC | H,D 295 | XP_005258940.1 | |
XM_011526938.1 | 1468 | Intron | XP_011525240.1 |