Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGACGGGCAGGGCTGCCACGGAGG[C/T]GGTGACCACGCTCTCAATGCCCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610396 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NKPD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NKPD1 - NTPase, KAP family P-loop domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRAPPC6A - trafficking protein particle complex 6A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270891.1 | 449 | Missense Mutation | ACC,GCC | T,A 142 | NP_001257820.1 | |
NM_001270892.1 | 449 | UTR 3 | NP_001257821.1 | |||
NM_001270893.1 | 449 | UTR 3 | NP_001257822.1 | |||
NM_024108.2 | 449 | Missense Mutation | ACC,GCC | T,A 156 | NP_077013.1 | |
XM_017027295.1 | 449 | UTR 3 | XP_016882784.1 |