Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCTGCTGGTCACAGCTCAGTGCG[C/T]GATGCCTTGGCCACAATCACCAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606746 MIM: 615869 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYDGF PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MYDGF - myeloid derived growth factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019107.3 | 576 | Silent Mutation | TCA,TCG | S,S 169 | NP_061980.1 | |
XM_017026987.1 | 576 | Intron | XP_016882476.1 |
TNFAIP8L1 - TNF alpha induced protein 8 like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |