Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGCCGCCGCCTCCTCCAGCACCT[C/T]GGAGAAGCGCTCGAAGTCGGCGTAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603289 | ||||||||||||||||||||
Literature Links: |
DAPK3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DAPK3 - death associated protein kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001348.2 | 1276 | Missense Mutation | AAG,GAG | K,E 327 | NP_001339.1 | |
XM_005259508.1 | 1276 | Missense Mutation | AAG,GAG | K,E 327 | XP_005259565.1 |
MIR637 - microRNA 637 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |