Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCCCTCCGAGACTGCCCCCGAAG[A/C]CCCTGGGGCCGGAGCAACTGTGACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610671 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NAT14 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NAT14 - N-acetyltransferase 14 (putative) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SSC5D - scavenger receptor cysteine rich family member with 5 domains | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144950.1 | 927 | Missense Mutation | AGA,AGC | R,S 280 | NP_001138422.1 | |
NM_001195267.1 | 927 | Missense Mutation | AGA,AGC | R,S 280 | NP_001182196.1 |
ZNF628 - zinc finger protein 628 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |