Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGACGTGTGTATCTCCATCCTCC[A/C]CCCGCCGGTGGACGACCCCCAGAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 116948 MIM: 600311 | ||||||||||||||||||||
Literature Links: |
CDC34 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDC34 - cell division cycle 34 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004359.1 | 492 | Missense Mutation | CAC,CCC | H,P 98 | NP_004350.1 | |
XM_005259690.3 | 492 | Missense Mutation | CAC,CCC | H,P 98 | XP_005259747.1 | |
XM_006722952.2 | 492 | Missense Mutation | CAC,CCC | H,P 98 | XP_006723015.1 |
GZMM - granzyme M | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |