Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAGGCGTAGCGGGAGTTGAGGAC[A/G]CGGGCAATGCCCTCTTCTGTGCTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600283 | ||||||||||||||||||||
Literature Links: |
GRIK5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GRIK5 - glutamate ionotropic receptor kainate type subunit 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301030.1 | 3585 | Silent Mutation | CGC,CGT | R,R 711 | NP_001287959.1 | |
NM_002088.4 | 3585 | Silent Mutation | CGC,CGT | R,R 711 | NP_002079.3 | |
XM_005258821.3 | 3585 | Silent Mutation | CGC,CGT | R,R 711 | XP_005258878.1 | |
XM_011526862.2 | 3585 | Silent Mutation | CGC,CGT | R,R 712 | XP_011525164.1 | |
XM_011526863.2 | 3585 | Silent Mutation | CGC,CGT | R,R 711 | XP_011525165.1 | |
XM_011526865.1 | 3585 | Silent Mutation | CGC,CGT | R,R 712 | XP_011525167.1 | |
XM_011526867.1 | 3585 | Silent Mutation | CGC,CGT | R,R 643 | XP_011525169.1 | |
XM_011526868.1 | 3585 | Silent Mutation | CGC,CGT | R,R 631 | XP_011525170.1 | |
XM_011526869.2 | 3585 | Intron | XP_011525171.1 | |||
XM_011526870.2 | 3585 | Intron | XP_011525172.1 | |||
XM_011526871.2 | 3585 | Intron | XP_011525173.1 | |||
XM_017026713.1 | 3585 | Silent Mutation | CGC,CGT | R,R 644 | XP_016882202.1 |