Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTCGCAGTAGTCCCAGTCGTGCC[A/G]CTCCTGCTTGCAGGCCAGGAAGTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603842 MIM: 610057 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NDUFB7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NDUFB7 - NADH:ubiquinone oxidoreductase subunit B7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004146.5 | 327 | Missense Mutation | CGG,TGG | R,W 84 | NP_004137.2 | |
XM_011528039.2 | 327 | Missense Mutation | CGG,TGG | R,W 46 | XP_011526341.1 |
TECR - trans-2,3-enoyl-CoA reductase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321170.1 | 327 | Intron | NP_001308099.1 | |||
NM_138501.5 | 327 | Intron | NP_612510.1 | |||
XM_006722945.1 | 327 | Intron | XP_006723008.1 | |||
XM_011528442.2 | 327 | Intron | XP_011526744.1 |