Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCGAGACTGGCTTCTGGGTGCTTC[A/G]GGTCCTCCGCTGACTCTGGAAGACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607382 MIM: 612536 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLIP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLIP3 - CAP-Gly domain containing linker protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC101927572 - uncharacterized LOC101927572 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290056.1 | 646 | Intron | NP_001276985.1 |
THAP8 - THAP domain containing 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152658.2 | 646 | Nonsense Mutation | CGA,TGA | R,* 98 | NP_689871.1 | |
XM_005258623.2 | 646 | Nonsense Mutation | CGA,TGA | R,* 55 | XP_005258680.1 | |
XM_011526604.2 | 646 | Nonsense Mutation | CGA,TGA | R,* 55 | XP_011524906.1 |