Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAGGTTCCTGTGGCTGCAGACTTG[C/G]TTACAGACCATGAGGAGGTAAGTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF419 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF419 - zinc finger protein 419 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098491.1 | 295 | Missense Mutation | CTT,GTT | L,V 19 | NP_001091961.1 | |
NM_001098492.1 | 295 | Intron | NP_001091962.1 | |||
NM_001098493.1 | 295 | Intron | NP_001091963.1 | |||
NM_001098494.1 | 295 | Missense Mutation | CTT,GTT | L,V 19 | NP_001091964.1 | |
NM_001098495.1 | 295 | Intron | NP_001091965.1 | |||
NM_001098496.1 | 295 | Intron | NP_001091966.1 | |||
NM_001291743.1 | 295 | Missense Mutation | CTT,GTT | L,V 19 | NP_001278672.1 | |
NM_001291744.1 | 295 | UTR 5 | NP_001278673.1 | |||
NM_001291745.1 | 295 | Intron | NP_001278674.1 | |||
NM_024691.3 | 295 | Missense Mutation | CTT,GTT | L,V 19 | NP_078967.3 |
ZNF772 - zinc finger protein 772 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF773 - zinc finger protein 773 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |