Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTGCTGGTGGAGCGGGAGCGGCC[C/T]CCCGGCCAGGTGCCCCTCATCATCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605770 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ILVBL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ILVBL - ilvB acetolactate synthase like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYDE1 - synapse defective Rho GTPase homolog 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300910.1 | 1297 | Silent Mutation | CCC,CCT | P,P 342 | NP_001287839.1 | |
NM_033025.5 | 1297 | Silent Mutation | CCC,CCT | P,P 409 | NP_149014.3 |