Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTGCACAGGTTGGTGTAGACACC[A/G]GGGTAGCCAGGCAGGGCGCAGCGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606135 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLK14 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KLK14 - kallikrein related peptidase 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001311182.1 | 925 | Silent Mutation | CCC,CCT | P,P 247 | NP_001298111.1 | |
NM_022046.5 | 925 | Silent Mutation | CCC,CCT | P,P 247 | NP_071329.2 | |
XM_017026820.1 | 925 | Intron | XP_016882309.1 |