Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCATGAGGTCGTGGCTGTAGTCCT[C/T]GTCTGCTTGGCGGGTGTGGTTCTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 147910 MIM: 610601 | ||||||||||||||||||||
Literature Links: |
KLK1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KLK1 - kallikrein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002257.3 | 1278 | Missense Mutation | AAG,GAG | K,E 115 | NP_002248.1 | |
XM_011526942.1 | 1278 | Missense Mutation | AAG,GAG | K,E 13 | XP_011525244.1 |
KLK15 - kallikrein related peptidase 15 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MGC45922 - uncharacterized LOC284365 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |