Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTACATTCAAAGGGTTTCTCACC[C/T]GTACAAGTTTTTGATGCAGAGAAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF780B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF780B - zinc finger protein 780B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005851.2 | 2708 | Missense Mutation | CAG,CGG | Q,R 804 | NP_001005851.1 | |
XM_005258590.4 | 2708 | Missense Mutation | CAG,CGG | Q,R 805 | XP_005258647.1 | |
XM_005258591.4 | 2708 | Missense Mutation | CAG,CGG | Q,R 805 | XP_005258648.1 | |
XM_005258592.3 | 2708 | Missense Mutation | CAG,CGG | Q,R 805 | XP_005258649.1 | |
XM_005258593.2 | 2708 | Missense Mutation | CAG,CGG | Q,R 804 | XP_005258650.1 | |
XM_005258595.3 | 2708 | Missense Mutation | CAG,CGG | Q,R 805 | XP_005258652.1 | |
XM_006723072.3 | 2708 | Missense Mutation | CAG,CGG | Q,R 805 | XP_006723135.1 | |
XM_011526593.2 | 2708 | Missense Mutation | CAG,CGG | Q,R 805 | XP_011524895.1 | |
XM_017026425.1 | 2708 | Missense Mutation | CAG,CGG | Q,R 804 | XP_016881914.1 | |
XM_017026426.1 | 2708 | Missense Mutation | CAG,CGG | Q,R 804 | XP_016881915.1 | |
XM_017026427.1 | 2708 | Missense Mutation | CAG,CGG | Q,R 804 | XP_016881916.1 |