Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTCCCCAACGTTGTTGGGCTCTC[C/T]TCTGTTCCAATACTGCTTGAAGCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604672 | ||||||||||||||||||||
Literature Links: |
CD209 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CD209 - CD209 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144893.1 | 720 | Missense Mutation | AGA,GGA | R,G 210 | NP_001138365.1 | |
NM_001144894.1 | 720 | Missense Mutation | AGA,GGA | R,G 302 | NP_001138366.1 | |
NM_001144895.1 | 720 | Missense Mutation | AGA,GGA | R,G 254 | NP_001138367.1 | |
NM_001144896.1 | 720 | Missense Mutation | AGA,GGA | R,G 322 | NP_001138368.1 | |
NM_001144897.1 | 720 | Missense Mutation | AGA,GGA | R,G 340 | NP_001138369.1 | |
NM_001144899.1 | 720 | Missense Mutation | AGA,GGA | R,G 185 | NP_001138371.1 | |
NM_021155.3 | 720 | Missense Mutation | AGA,GGA | R,G 346 | NP_066978.1 | |
XM_005272472.4 | 720 | Missense Mutation | AGA,GGA | R,G 208 | XP_005272529.1 |