Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACCTGGATCCTCTGCAGTCCCCC[C/T]GAGTCCAGTGCCCACCGTCCCGGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 126350 MIM: 608801 MIM: 600599 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNASE2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNASE2 - deoxyribonuclease II, lysosomal | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GCDH - glutaryl-CoA dehydrogenase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KLF1 - Kruppel like factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011527642.2 | 818 | Missense Mutation | XP_011525944.1 |