Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGTCGTTGGGCCTCTGGTTGTGCT[C/T]AGAAGTGGCTGAGGCCGCTGTGCCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605072 MIM: 601032 MIM: 176802 | ||||||||||||||||||||
Literature Links: |
GIPC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GIPC1 - GIPC PDZ domain containing family member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PKN1 - protein kinase N1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002741.3 | 1326 | Intron | NP_002732.3 | |||
NM_213560.1 | 1326 | Intron | NP_998725.1 | |||
XM_011528128.1 | 1326 | Intron | XP_011526430.1 |
PTGER1 - prostaglandin E receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000955.2 | 1326 | Silent Mutation | TAA,TGA | *,* 403 | NP_000946.2 |