Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCCCTGCAGTCAAGGCGCGGCGC[A/G]GCCCCCGTGCCGCTTCCGGTGCTCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100419840 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC100419840 - zinc finger protein 446 pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105372483 - uncharacterized LOC105372483 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF497 - zinc finger protein 497 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF837 - zinc finger protein 837 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138466.1 | 2571 | Missense Mutation | CGC,TGC | R,C 528 | NP_612475.1 | |
XM_017026242.1 | 2571 | Missense Mutation | CGC,TGC | R,C 528 | XP_016881731.1 |