Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGGATGGCCGGCTCCTCTCCACC[A/G]TCATCCGTGCCTTGGACACACCGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613332 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MARCH2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MARCH2 - membrane associated ring-CH-type finger 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005415.1 | 321 | Missense Mutation | ATC,GTC | I,V 51 | NP_001005415.1 | |
NM_001005416.1 | 321 | Missense Mutation | ATC,GTC | I,V 51 | NP_001005416.1 | |
NM_016496.4 | 321 | Missense Mutation | ATC,GTC | I,V 51 | NP_057580.3 | |
XM_006722763.3 | 321 | Missense Mutation | ATC,GTC | I,V 51 | XP_006722826.1 | |
XM_011528046.2 | 321 | Missense Mutation | ATC,GTC | I,V 51 | XP_011526348.1 | |
XM_011528047.2 | 321 | Missense Mutation | ATC,GTC | I,V 51 | XP_011526349.1 | |
XM_017026853.1 | 321 | Missense Mutation | ATC,GTC | I,V 51 | XP_016882342.1 | |
XM_017026854.1 | 321 | Silent Mutation | CCA,CCG | P,P 7 | XP_016882343.1 |