Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGTCCAGCCAGAAGGTCCACCCT[A/G]GTGAGACATGTGGCCCACCCTTGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF548 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF548 - zinc finger protein 548 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172773.1 | 542 | Missense Mutation | AGT,GGT | S,G 98 | NP_001166244.1 | |
NM_152909.3 | 542 | Missense Mutation | AGT,GGT | S,G 86 | NP_690873.2 | |
XM_017026316.1 | 542 | Missense Mutation | AGT,GGT | S,G 89 | XP_016881805.1 |