Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCAGTATGACATCTCATATGAAC[C/T]GAAAGGTATCCACCGTAATTAAAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF841 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF841 - zinc finger protein 841 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136499.1 | 3726 | Silent Mutation | TCA,TCG | S,S 557 | NP_001129971.1 | |
NM_001321349.1 | 3726 | Silent Mutation | TCA,TCG | S,S 441 | NP_001308278.1 | |
XM_011526833.2 | 3726 | Intron | XP_011525135.1 | |||
XM_017026652.1 | 3726 | Intron | XP_016882141.1 | |||
XM_017026653.1 | 3726 | Silent Mutation | TCA,TCG | S,S 558 | XP_016882142.1 | |
XM_017026654.1 | 3726 | Silent Mutation | TCA,TCG | S,S 558 | XP_016882143.1 | |
XM_017026655.1 | 3726 | Intron | XP_016882144.1 | |||
XM_017026656.1 | 3726 | Silent Mutation | TCA,TCG | S,S 557 | XP_016882145.1 | |
XM_017026657.1 | 3726 | Intron | XP_016882146.1 | |||
XM_017026658.1 | 3726 | Intron | XP_016882147.1 | |||
XM_017026659.1 | 3726 | Silent Mutation | TCA,TCG | S,S 559 | XP_016882148.1 | |
XM_017026660.1 | 3726 | Intron | XP_016882149.1 | |||
XM_017026661.1 | 3726 | Intron | XP_016882150.1 | |||
XM_017026662.1 | 3726 | Silent Mutation | TCA,TCG | S,S 557 | XP_016882151.1 | |
XM_017026663.1 | 3726 | Intron | XP_016882152.1 |