Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCTGGGCGCGCTGCCCAGCAAGA[A/T]GCGGACCTGGGTGCAGTCACAGGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZSCAN1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZSCAN1 - zinc finger and SCAN domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182572.3 | 2141 | Missense Mutation | AAG,ATG | K,M 94 | NP_872378.3 | |
XM_006723149.2 | 2141 | Missense Mutation | AAG,ATG | K,M 134 | XP_006723212.1 | |
XM_017026609.1 | 2141 | Missense Mutation | AAG,ATG | K,M 134 | XP_016882098.1 | |
XM_017026610.1 | 2141 | Missense Mutation | AAG,ATG | K,M 94 | XP_016882099.1 |