Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGATGAGAGGACAGAATAAAGCTT[C/T]TGCCACATTCTTCACACTTATAGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF708 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF708 - zinc finger protein 708 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297560.1 | 1463 | Intron | NP_001284489.1 | |||
NM_001297561.1 | 1463 | Intron | NP_001284490.1 | |||
NM_021269.2 | 1463 | Intron | NP_067092.2 | |||
XM_017027201.1 | 1463 | Intron | XP_016882690.1 | |||
XM_017027202.1 | 1463 | Missense Mutation | AAA,AGA | K,R 407 | XP_016882691.1 | |
XM_017027203.1 | 1463 | Missense Mutation | AAA,AGA | K,R 407 | XP_016882692.1 | |
XM_017027204.1 | 1463 | Missense Mutation | AAA,AGA | K,R 483 | XP_016882693.1 |