Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCTGGACCCAGCTCAGAGGAATC[G/T]CTACAGAGACGTGATGCTGGAAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603996 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF114 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF114 - zinc finger protein 114 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301062.1 | 438 | UTR 5 | NP_001287991.1 | |||
NM_153608.2 | 438 | Missense Mutation | CGC,CTC | R,L 30 | NP_705836.1 | |
XM_011526574.2 | 438 | Missense Mutation | CGC,CTC | R,L 76 | XP_011524876.1 | |
XM_017026414.1 | 438 | Missense Mutation | CGC,CTC | R,L 76 | XP_016881903.1 | |
XM_017026415.1 | 438 | Missense Mutation | CGC,CTC | R,L 30 | XP_016881904.1 |