Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGACATTGATTCGCTGGTTCTG[A/G]CTGTTGGTTCCTGGGAGAATCACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611048 MIM: 602309 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP1R15A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPP1R15A - protein phosphatase 1 regulatory subunit 15A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TULP2 - tubby like protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003323.2 | 1337 | Silent Mutation | AGC,AGT | S,S 414 | NP_003314.2 | |
XM_006723347.3 | 1337 | Silent Mutation | AGC,AGT | S,S 414 | XP_006723410.1 | |
XM_006723348.3 | 1337 | Silent Mutation | AGC,AGT | S,S 414 | XP_006723411.1 | |
XM_011527252.2 | 1337 | Silent Mutation | AGC,AGT | S,S 414 | XP_011525554.1 | |
XM_011527253.2 | 1337 | Silent Mutation | AGC,AGT | S,S 411 | XP_011525555.1 | |
XM_011527254.2 | 1337 | Intron | XP_011525556.1 | |||
XM_011527255.2 | 1337 | Silent Mutation | AGC,AGT | S,S 370 | XP_011525557.1 | |
XM_011527256.2 | 1337 | Silent Mutation | AGC,AGT | S,S 370 | XP_011525558.1 |