Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGAAGTCCCCGCTCTGGATGCCG[C/T]GTGCTGTCGAGCGGCCAGCATTGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610477 | ||||||||||||||||||||
Literature Links: |
TMPRSS9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMPRSS9 - transmembrane protease, serine 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011527978.2 | 332 | Missense Mutation | GCG,GTG | A,V 24 | XP_011526280.1 | |
XM_011527980.1 | 332 | Intron | XP_011526282.1 | |||
XM_011527982.2 | 332 | Intron | XP_011526284.1 |