Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGACCTCCAGAAGGAACTGAGTGA[C/T]ATATGGTGATGCCCAGCCTGCAGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613771 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC159 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC159 - coiled-coil domain containing 159 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080503.2 | 955 | Silent Mutation | GAC,GAT | D,D 228 | NP_001073972.2 | |
XM_006722643.1 | 955 | Silent Mutation | GAC,GAT | D,D 264 | XP_006722706.1 | |
XM_011527678.2 | 955 | Silent Mutation | GAC,GAT | D,D 227 | XP_011525980.2 | |
XM_017026255.1 | 955 | Silent Mutation | GAC,GAT | D,D 264 | XP_016881744.1 | |
XM_017026256.1 | 955 | Silent Mutation | GAC,GAT | D,D 262 | XP_016881745.1 | |
XM_017026257.1 | 955 | Silent Mutation | GAC,GAT | D,D 237 | XP_016881746.1 | |
XM_017026258.1 | 955 | Silent Mutation | GAC,GAT | D,D 228 | XP_016881747.1 | |
XM_017026259.1 | 955 | Silent Mutation | GAC,GAT | D,D 227 | XP_016881748.1 | |
XM_017026260.1 | 955 | Silent Mutation | GAC,GAT | D,D 227 | XP_016881749.1 | |
XM_017026261.1 | 955 | Silent Mutation | GAC,GAT | D,D 205 | XP_016881750.1 | |
XM_017026262.1 | 955 | Silent Mutation | GAC,GAT | D,D 205 | XP_016881751.1 | |
XM_017026263.1 | 955 | Silent Mutation | GAC,GAT | D,D 205 | XP_016881752.1 | |
XM_017026264.1 | 955 | Silent Mutation | GAC,GAT | D,D 205 | XP_016881753.1 |
PLPPR2 - phospholipid phosphatase related 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM205 - transmembrane protein 205 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |