Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGGTAAGTTGTGAACTACGAATAA[A/G]GGCCATTCCACACTCCCTGCATTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614275 | ||||||||||||||||||||
Literature Links: |
ZNF565 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF565 - zinc finger protein 565 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042474.2 | 2171 | Missense Mutation | CTT,TTT | L,F 456 | NP_001035939.1 | |
NM_152477.4 | 2171 | Missense Mutation | CTT,TTT | L,F 456 | NP_689690.3 | |
XM_011526512.2 | 2171 | Missense Mutation | CTT,TTT | L,F 456 | XP_011524814.1 | |
XM_011526514.2 | 2171 | Missense Mutation | CTT,TTT | L,F 456 | XP_011524816.1 | |
XM_017026341.1 | 2171 | Missense Mutation | CTT,TTT | L,F 456 | XP_016881830.1 | |
XM_017026342.1 | 2171 | Missense Mutation | CTT,TTT | L,F 391 | XP_016881831.1 | |
XM_017026343.1 | 2171 | Missense Mutation | CTT,TTT | L,F 391 | XP_016881832.1 |