Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCCTCAGTGGTGCCCGACGGGGG[A/T]TCGGCAAGGGCGTCCCCAGGGGGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZSCAN18 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZSCAN18 - zinc finger and SCAN domain containing 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145542.1 | 1146 | Missense Mutation | GAA,GAT | E,D 372 | NP_001139014.1 | |
NM_001145543.1 | 1146 | Missense Mutation | GAA,GAT | E,D 316 | NP_001139015.1 | |
NM_001145544.1 | 1146 | Missense Mutation | GAA,GAT | E,D 180 | NP_001139016.1 | |
NM_023926.4 | 1146 | Missense Mutation | GAA,GAT | E,D 316 | NP_076415.3 | |
XM_005259174.4 | 1146 | Missense Mutation | GAA,GAT | E,D 316 | XP_005259231.1 | |
XM_006723335.2 | 1146 | Missense Mutation | GAA,GAT | E,D 316 | XP_006723398.1 | |
XM_011527237.2 | 1146 | Missense Mutation | GAA,GAT | E,D 371 | XP_011525539.1 | |
XM_011527238.1 | 1146 | Missense Mutation | GAA,GAT | E,D 316 | XP_011525540.1 | |
XM_011527239.2 | 1146 | Missense Mutation | GAA,GAT | E,D 316 | XP_011525541.1 | |
XM_017027169.1 | 1146 | Missense Mutation | GAA,GAT | E,D 315 | XP_016882658.1 | |
XM_017027170.1 | 1146 | Missense Mutation | GAA,GAT | E,D 315 | XP_016882659.1 | |
XM_017027171.1 | 1146 | Missense Mutation | GAA,GAT | E,D 315 | XP_016882660.1 |