Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAACTACAGGAACGTGGTCTCCCTG[C/G]GTAAGGCAAGCATCACTAATTCCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF554 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF554 - zinc finger protein 554 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102651.1 | 1418 | Missense Mutation | CAA,GAA | Q,E 85 | NP_001096121.1 | |
XM_006722635.3 | 1418 | UTR 5 | XP_006722698.1 | |||
XM_011527660.2 | 1418 | Missense Mutation | CAA,GAA | Q,E 34 | XP_011525962.1 | |
XM_011527662.2 | 1418 | Missense Mutation | CAA,GAA | Q,E 34 | XP_011525964.1 | |
XM_011527664.2 | 1418 | UTR 5 | XP_011525966.1 | |||
XM_011527667.2 | 1418 | Missense Mutation | CAT,GAT | H,D 85 | XP_011525969.1 | |
XM_017026234.1 | 1418 | Missense Mutation | CCC,GCC | P,A 85 | XP_016881723.1 | |
XM_017026235.1 | 1418 | UTR 5 | XP_016881724.1 | |||
XM_017026236.1 | 1418 | Intron | XP_016881725.1 |