Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTTTTTCTTTGGCACACACGAAA[C/T]GTAAGTGTCCCCTTCTGGGGCTTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
HDGFRP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HDGFRP2 - hepatoma-derived growth factor-related protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001520.2 | 664 | Missense Mutation | ACA,ATA | T,I 50 | NP_001001520.1 | |
NM_032631.3 | 664 | Missense Mutation | ACA,ATA | T,I 50 | NP_116020.1 | |
XM_011528375.2 | 664 | Missense Mutation | ACA,ATA | T,I 74 | XP_011526677.1 | |
XM_011528376.2 | 664 | Missense Mutation | ACA,ATA | T,I 74 | XP_011526678.1 | |
XM_011528377.2 | 664 | Missense Mutation | ACA,ATA | T,I 74 | XP_011526679.1 |