Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTAGGTTTGAATGTACACTAAATG[C/G]TTTGCCGCACTCATTACACTTATAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF347 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF347 - zinc finger protein 347 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF665 - zinc finger protein 665 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024733.3 | 1937 | Missense Mutation | CCA,GCA | P,A 578 | NP_079009.3 | |
XM_005259266.4 | 1937 | Missense Mutation | CCA,GCA | P,A 578 | XP_005259323.1 | |
XM_011527325.2 | 1937 | Missense Mutation | CCA,GCA | P,A 595 | XP_011525627.1 | |
XM_011527327.2 | 1937 | Missense Mutation | CCA,GCA | P,A 576 | XP_011525629.1 |