Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGCGGTCAGCACTGACTACTGGCT[G/T]TACATGGAAGAAGGCACAGTGCTAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606899 MIM: 176980 | ||||||||||||||||||||
Literature Links: |
CACNG7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CACNG7 - calcium voltage-gated channel auxiliary subunit gamma 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031896.4 | 224 | Silent Mutation | CTG,CTT | L,L 34 | NP_114102.2 | |
XM_005259124.3 | 224 | Silent Mutation | CTG,CTT | L,L 34 | XP_005259181.1 | |
XM_017027093.1 | 224 | Silent Mutation | CTG,CTT | L,L 34 | XP_016882582.1 |
PRKCG - protein kinase C gamma | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |