Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAGAGCTCACAGAGGCTGTGCCGC[A/G]CCCGCCCCACTGGCTGCTGGAGCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613247 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HDGFRP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HDGFRP2 - hepatoma-derived growth factor-related protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLIN4 - perilipin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080400.1 | 5833 | Missense Mutation | GCG,GTG | A,V 1291 | NP_001073869.1 | |
XM_006722866.2 | 5833 | Missense Mutation | GCG,GTG | A,V 1306 | XP_006722929.1 | |
XM_006722868.3 | 5833 | Missense Mutation | GCG,GTG | A,V 1305 | XP_006722931.1 | |
XM_011528233.2 | 5833 | Missense Mutation | GCG,GTG | A,V 1366 | XP_011526535.1 | |
XM_017027192.1 | 5833 | Missense Mutation | GCG,GTG | A,V 1367 | XP_016882681.1 | |
XM_017027193.1 | 5833 | Missense Mutation | GCG,GTG | A,V 1367 | XP_016882682.1 | |
XM_017027194.1 | 5833 | Missense Mutation | GCG,GTG | A,V 1367 | XP_016882683.1 |