Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACGTGGGTGAGAAAGTGTACTGCC[A/C]GGAGGACTTCCTGGTGAGTCCAAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614790 | ||||||||||||||||||||
Literature Links: |
WTIP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
WTIP - Wilms tumor 1 interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080436.1 | 258 | Missense Mutation | CAG,CCG | Q,P 275 | NP_001073905.1 | |
XM_006723014.3 | 258 | Missense Mutation | CAG,CCG | Q,P 275 | XP_006723077.1 | |
XM_011526452.2 | 258 | Missense Mutation | CAG,CCG | Q,P 275 | XP_011524754.1 | |
XM_011526453.2 | 258 | Missense Mutation | CAG,CCG | Q,P 34 | XP_011524755.1 |