Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGAAACGCTCCGTTGGAGAATGC[C/T]GTTGAGATGAGTCCCTTCTTATCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606091 | ||||||||||||||||||||
Literature Links: |
LOC100129083 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC100129083 - uncharacterized LOC100129083 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SIGLEC10 - sialic acid binding Ig like lectin 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171156.1 | 1401 | Silent Mutation | ACA,ACG | T,T 488 | NP_001164627.1 | |
NM_001171157.1 | 1401 | Intron | NP_001164628.1 | |||
NM_001171158.1 | 1401 | Silent Mutation | ACA,ACG | T,T 403 | NP_001164629.1 | |
NM_001171159.1 | 1401 | Silent Mutation | ACA,ACG | T,T 393 | NP_001164630.1 | |
NM_001171161.1 | 1401 | Silent Mutation | ACA,ACG | T,T 303 | NP_001164632.1 | |
NM_001322105.1 | 1401 | Silent Mutation | ACA,ACG | T,T 361 | NP_001309034.1 | |
NM_033130.4 | 1401 | Silent Mutation | ACA,ACG | T,T 546 | NP_149121.2 | |
XM_005259366.4 | 1401 | Silent Mutation | ACA,ACG | T,T 456 | XP_005259423.1 | |
XM_005259367.3 | 1401 | Silent Mutation | ACA,ACG | T,T 398 | XP_005259424.1 | |
XM_011527439.1 | 1401 | Silent Mutation | ACA,ACG | T,T 498 | XP_011525741.1 |